1,2-Dibromobenzene
Catalog # EHERC12216710
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:0,1 g
- Seq:3020
Specifications
About this item
:
:235.91 g/mol
Boiling Pt:224 °C (1013 hPa)
Density:1.956 g/cm³ (20 °C)
Storage Temperature:Refrigerator
EINECS:209-507-3
UN:3082
ADR:9,III
CAS Number:583-53-9
Formula:C₆H₄Br₂
MDL Number:MFCD00000057