Order Entry
Austria
ContactUsLinkComponent
 
9,10-Diphenylanthracene for synthesis, Sigma-Aldrich®
 :  Merck
 
9,10-Diphenylanthracene for synthesis, Sigma-Aldrich®
 :  Merck

 

  • Solubility:
    (20 °C) insoluble
  • Melting Point:
    247 - 252 °C
  • Molar Mass:
    330.43 g/mol

 

Frequently Bought Together