ContactUsLinkComponent
PVF fluoropolymer gas and liquid sampling bags
PVF fluoropolymer gas and liquid sampling bags
Catalog # 300-0280
undefined
PVF fluoropolymer gas and liquid sampling bags
Catalog # 300-0280
Specifications
  • Packed:
    1 bag/pack
  • Capacity:
    3,8 L
  • Description:
    Gas sampling bags with on/off valve type
  • Seq:
    02
  • Pk:
    1
  • W×L:
    304,8×304,8 mm
Specifications
About this item
Made from 2 mil thick PVF fluoropolymer film. When the chemical inertness of FEP is not required, Chemware® heat-sealed, PVF gas sampling bags are the ideal choice. These reusable bags are the perfect economical alternative for analysis of non-corrosive gases. Can be used when the user is dealing with carbon monoxide, sulphur dioxide, hydrogen sulfide, radon and mercaptans. Ideal for extracting air samples from smoke stack exhaust systems in fuel gas sampling applications.
  • Tough, easy to use, reliable
  • Highly flexible – easy to handle and store
  • Leak proof, heat sealed all four sides
  • No aging – unlimited storage life
  • Unique seam construction for added strength
  • Non-adhesive, nothing sticks
Just as in Chemware®’s FEP bags, Chemware®’s PVF gas sampling bags accept two fitting options – either a nickel-plated brass on/off valve, or a PTFE/silicone septum fitting in a polypropylene housing. Durable, heat-sealed seams ensure leak-proof operation.