Precolumn filters, Upchurch Scientific®
Lieferant: UPCHURCH SCIENTIFIC
Zertifikate
Über diesen Artikel
These filters are ideal for higher pressure applications and to help prevent particulate contamination.
- For ¹/₁₆" Ø tubing
Frequently Bought Together