Test tubes with standard ground joints, conical bottom
Supplier: LENZ LABORGLAS GMBH & CO.KG
Certificates
About this item
Centrifuge tubes, made of DURAN® borosilicate glass 3.3.
- Excellent chemical resistance
- High temperature resistance
- Minimal thermal expansion giving relatively high resistance to temperature changes
Frequently Bought Together