Centrifuge tubes, ASTM
Supplier: LENZ LABORGLAS GMBH & CO.KG
Certificates
About this item
DURAN®, borosilicate glass.
- Withstands forces up to 700 ×g
- Good temperature resistance
- Good chemical resistance
With conical bottom or pear shaped.
Frequently Bought Together