


2-Pentyne ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:627-21-4
:
Formula:C₅H₈
MDL Number:MFCD00009380
:68.12 g/mol
Boiling Pt:55…56 °C
Melting Pt:–109 °C
Density:0.710
Flash Pt:–20 °C (–4 °F)
Storage Temperature:Ambient
EINECS:210-989-2
UN:3295
ADR:3,II
:GHS02
:GHS08
:GHS07
Related Information
Frequently Bought Together