Supelco® Organic Reference Standard, 1-Pentene
Supplier: Merck
Certificates
About this item
- GHS signal: GHS02, GHS08, GHS07
Caution: P210 - P233 - P301 + P310 - P303 + P361 + P353 - P331 - P403 + P233
Specifications
- CAS No.:109-67-1
- Storage conditions:2…8 °C
- Hazard class description:H224 - H304 - H315 - H319
- Shipping conditions:STD
- MDL No.:MFCD00003567
- M-Clarity quality level:MQ100
- Formula:CH₃CH₂CH₂CH=CH₂
- Molecular weight:70,13
- Density:0,641 g/ml at 25 °C (lit.)
- Flash point:–51 °C – Closed cup
- Boiling point:29.9 - 30,1 °C (lit.)
Specifications
Frequently Bought Together