Order Entry
Austria
ContactUsLinkComponent
1-Eicosene
1-Eicosene
Catalog # EHERC13113300
Supplier:  EHRENSTORFER
CAS Number:  
1-Eicosene
Catalog # EHERC13113300
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    0,1 g
  • Seq:
    3020

Specifications

About this item

 :
 :280.54 g/mol
Boiling Pt:342 °C (1013 hPa)
Melting Pt:60…62 °C
Density:0.795 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:222-374-6
CAS Number:3452-07-1
Formula:C₂₀H₄₀
MDL Number:MFCD00009013