Orders LinkContactUsLinkComponent
Volumetric flasks
Volumetric flasks
Supplier:   Brand
Volumetric flasks
Supplier:  Brand
Certificates
Volumetric flasks
Volumetric flasks
8 Options Available Below
Product Details & Documents
PFA, translucent, with PFA screw cap.
  • Inclusive one batch certificate
  • Exposure to temperatures up to 121 ºC (autoclaving) will not cause permanently exceeded error limits
  • Cleaning up to max. 60 °C is recommended to preserve marks and inscriptions
Error limits according to class A, DIN EN ISO 1042. Calibrated to contain (TC, In). PFA screw cap with molded sealing ring. Autoclavable, easy to clean.
*Wide neck.
(Height = Height without stopper).
Product Family Options
Spare screw caps for volumetric flasks
Product Information
  • Capacity
    Thread
    Height
    Tolerance
    Availability
    Price
  • GL 18
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 15,40 €
    15,40 €
    1 * 1 ST
       15,40 €
  • GL 25
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 19,60 €
    19,60 €
    1 * 1 ST
       19,60 €
Product Information
  • Capacity
    Thread
    Height
    Tolerance
    Availability
    Price
  • 25 ml
    GL 18
    108 mm
    0,04 ± ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 161,00 €
    161,00 €
    1 * 1 ST
       161,00 €
  • 50 ml
    GL 18
    143 mm
    0,06 ± ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 165,00 €
    165,00 €
    1 * 1 ST
       165,00 €
  • 100 ml
    GL 18
    166 mm
    0,10 ± ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 204,00 €
    204,00 €
    1 * 1 ST
       204,00 €
  • 250 ml
    GL 25
    222 mm
    0,15 ± ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 255,00 €
    255,00 €
    1 * 1 ST
       255,00 €
  • 500 ml
    GL 25
    262 mm
    0,25 ± ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 293,00 €
    293,00 €
    1 * 1 ST
       293,00 €
  • 10* ml
    GL 18
    90 mm
    0,04 ± ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 156,00 €
    156,00 €
    1 * 1 ST
       156,00 €