Adapters, delivery, straight, with drain tip
Supplier: LENZ LABORGLAS GMBH & CO.KG
Certificates
About this item
DURAN®, borosilicate glass.
- Very good chemical resistance
- High temperature resistance
- Minimal thermal expansion giving relatively high resistance to temperature changes
Delivery adapters, straight, with NS socket and NS cone, with drain tip.
Frequently Bought Together