Measuring cylinder, tall form, PP
Supplier: HIRSCHMANN
Certificates
About this item
PP, translucent, tall form, class B.
- Calibrated to 'In'
- Hexagonal base, ring mark on the principal points
- Thermal stresses of up to +80 °C do not result in the tolerance limit being permanently exceeded
Certifications: DIN 12681 B / ISO 6706
Frequently Bought Together