VWR® Traceable® K-Type Probes
Supplier: VWR Collection
Jump to:
About this item
2 Options Available Below
Product Details & Documents
Documents
Product Family Options
Product Information
- RangeØ×LAvailabilityPrice
- 620-2005Stainless steel probe 4014, with 1 m cableStainless steel probe 4014, with 1 m cable−50…+700 °C3,30×190,5 mmLog in for availability1 * 1 ST101,00 €
- 620-2012K-type fast response probe 4028, with 1,2 m coated cableK-type fast response probe 4028, with 1,2 m coated cable–40…+250 °C15×1219 mmLog in for availability1 * 1 ST52,50 €