Condensers, Allihn, for Soxleth extractors
Supplier: LENZ LABORGLAS GMBH & CO.KG
Certificates
About this item
DURAN® borosilicate glass 3.3.
- Excellent chemical resistance
- High temperature resistance
- Minimal thermal expansion giving relatively high resistance to temperature changes
For condensing vapours. With NS cone. Water connection with glass nozzles.
Frequently Bought Together