Distillation columns, Vigreux, with fixed vacuum jacket
Supplier: LENZ LABORGLAS GMBH & CO.KG
Certificates
About this item
DURAN®, borosilicate glass.
- Excellent chemical resistance
- High temperature resistance
- Minimal thermal expansion giving relatively high resistance to temperature changes
With NS socket, NS cone and with fixed vaccum jacket.
Frequently Bought Together