Distillation columns, Hempel, with fixed vacuum jacket
Supplier: LENZ LABORGLAS GMBH & CO.KG
Certificates
About this item
DURAN®, borosilicate glass.
- Excellent chemical resistance
- High temperature resistance
- Minimal thermal expansion giving relatively high resistance to temperature changes
For packings like Raschig rings or glass beads, with NS socket and NS cone, with fixed vacuum jacket.
Frequently Bought Together