

2-Methyloctane, Sigma-Aldrich®
Catalog # SIAL68170-5ML
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:5 mL
- Seq:2210
Specifications
About this item
:
:128.26 g/mol
Boiling Pt:143 °C (1013 hPa)
Melting Pt:–97 °C
Density:0.7095 g/cm³ (25 °C)
Storage Temperature:Ambient
EINECS:221-748-6
UN:1920
ADR:3,III
:GHS02
:GHS08
CAS Number:3221-61-2
Formula:C₉H₂₀
MDL Number:MFCD00027307