Distilling bridges with splash head
Supplier: LENZ LABORGLAS GMBH & CO.KG
Certificates
About this item
DURAN®, borosilicate glass 3.3.
- Excellent chemical resistance
- High temperature resistance
- Minimal thermal expansion, giving relatively high resistance to temperature changes
With two vertical NS cones and splash head.
Frequently Bought Together