Order Entry
Austria
Orders LinkContactUsLinkComponent
Buffer solutions

Buffer solutions

Buffer solutions
Supplier:  REAGECON
Certificates
Buffer solutions
Buffer solutions

82 Options Available Below

Product Details & Documents

 

Product Family Options

pH buffers

Product Information

  • pH value
    pH accuracy
    Colour
    Reference temperature
    Pack type
    Pk
    Availability
    Price
  • Buffer solution
    1,00
    ±0,02
    20 °C
    Plastic bottle
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL - 31,90 €
    31,90 €
    1 * 500 mL
       31,90 €

  • REGE1010525
    Buffer solution
    Buffer solution
    1,00
    ±0,02
    25 °C
    Plastic bottle
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL - 32,90 €
    32,90 €
    1 * 500 mL
       32,90 €

  • REGE1010
    Buffer solution
    Buffer solution
    1,00
    ±0,02
    20 °C
    Plastic bottle
    1 L
    Log in for availability
    Log in for availability
    1 * 1 L
    1 * 1 L - 46,90 €
    46,90 €
    1 * 1 L
       46,90 €

  • REGE101025
    Buffer solution
    Buffer solution
    1,00
    ±0,02
    25 °C
    Plastic bottle
    1 L
    Log in for availability
    Log in for availability
    1 * 1 L
    1 * 1 L - 49,30 €
    49,30 €
    1 * 1 L
       49,30 €

  • REGE5010
    Buffer solution
    Buffer solution
    1,00
    ±0,02
    20 °C
    Plastic bottle
    5 L
    Log in for availability
    Log in for availability
    1 * 5 L
    1 * 5 L - 141,00 €
    141,00 €
    1 * 5 L
       141,00 €

  • Buffer solution
    2,00
    ±0,02
    20 °C
    Plastic bottle
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL - 31,90 €
    31,90 €
    1 * 500 mL
       31,90 €

  • Buffer solution
    2,00
    ±0,02
    25 °C
    Plastic bottle
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL - 32,90 €
    32,90 €
    1 * 500 mL
       32,90 €

  • Buffer solution
    2,00
    ±0,02
    20 °C
    Plastic bottle
    1 L
    Log in for availability
    Log in for availability
    1 * 1 L
    1 * 1 L - 46,90 €
    46,90 €
    1 * 1 L
       46,90 €

  • REGE102025
    Buffer solution
    Buffer solution
    2,00
    ±0,02
    25 °C
    Plastic bottle
    1 L
    Log in for availability
    Log in for availability
    1 * 1 L
    1 * 1 L - 49,30 €
    49,30 €
    1 * 1 L
       49,30 €

  • REGE5020
    Buffer solution
    Buffer solution
    2,00
    ±0,02
    20 °C
    Plastic bottle
    5 L
    Log in for availability
    Log in for availability
    1 * 5 L
    1 * 5 L - 141,00 €
    141,00 €
    1 * 5 L
       141,00 €

  • REGE10305
    Buffer solution
    Buffer solution
    3,00
    ±0,02
    20 °C
    Plastic bottle
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL - 31,90 €
    31,90 €
    1 * 500 mL
       31,90 €

  • REGE1030525
    Buffer solution
    Buffer solution
    3,00
    ±0,02
    25 °C
    Plastic bottle
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL - 32,90 €
    32,90 €
    1 * 500 mL
       32,90 €

  • REGE1030
    Buffer solution
    Buffer solution
    3,00
    ±0,02
    20 °C
    Plastic bottle
    1 L
    Log in for availability
    Log in for availability
    1 * 1 L
    1 * 1 L - 46,90 €
    46,90 €
    1 * 1 L
       46,90 €

  • Buffer solution
    3,00
    ±0,02
    25 °C
    Plastic bottle
    1 L
    Log in for availability
    Log in for availability
    1 * 1 L
    1 * 1 L - 49,30 €
    49,30 €
    1 * 1 L
       49,30 €

  • REGE5030
    Buffer solution
    Buffer solution
    3,00
    ±0,02
    20 °C
    Plastic bottle
    5 L
    Log in for availability
    Log in for availability
    1 * 5 L
    1 * 5 L - 141,00 €
    141,00 €
    1 * 5 L
       141,00 €

  • Buffer solution
    4,00
    ±0,01
    20 °C
    Plastic bottle
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL - 30,50 €
    30,50 €
    1 * 500 mL
       30,50 €

  • Buffer solution
    4,00
    ±0,01
    25 °C
    Plastic bottle
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL - 31,90 €
    31,90 €
    1 * 500 mL
       31,90 €

  • REGECC1040525
    Buffer solution
    Buffer solution
    4,00
    ±0,01
    25 °C
    Plastic bottle
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL - 32,90 €
    32,90 €
    1 * 500 mL
       32,90 €

  • Buffer solution
    4,00
    ±0,01
    20 °C
    Plastic bottle
    1 L
    Log in for availability
    Log in for availability
    1 * 1 L
    1 * 1 L - 43,50 €
    43,50 €
    1 * 1 L
       43,50 €

  • Buffer solution
    4,00
    ±0,01
    25 °C
    Plastic bottle
    1 L
    Log in for availability
    Log in for availability
    1 * 1 L
    1 * 1 L - 46,20 €
    46,20 €
    1 * 1 L
       46,20 €

  • REGECC104025
    Buffer solution
    Buffer solution
    4,00
    ±0,01
    25 °C
    Plastic bottle
    1 L
    Log in for availability
    Log in for availability
    1 * 1 L
    1 * 1 L - 69,50 €
    69,50 €
    1 * 1 L
       69,50 €

  • REGE5040
    Buffer solution
    Buffer solution
    4,00
    ±0,01
    20 °C
    Plastic bottle
    5 L
    Log in for availability
    Log in for availability
    1 * 5 L
    1 * 5 L - 127,00 €
    127,00 €
    1 * 5 L
       127,00 €

  • REGE504025
    Buffer solution
    Buffer solution
    4,00
    ±0,01
    25 °C
    Plastic bottle
    5 L
    Log in for availability
    Log in for availability
    1 * 5 L
    1 * 5 L - 187,00 €
    187,00 €
    1 * 5 L
       187,00 €

  • Buffer solution
    5,00
    ±0,01
    20 °C
    Plastic bottle
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL - 31,90 €
    31,90 €
    1 * 500 mL
       31,90 €

  • REGE1050525
    Buffer solution
    Buffer solution
    5,00
    ±0,01
    25 °C
    Plastic bottle
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL - 32,90 €
    32,90 €
    1 * 500 mL
       32,90 €

  • REGE1050
    Buffer solution
    Buffer solution
    5,00
    ±0,01
    20 °C
    Plastic bottle
    1 L
    Log in for availability
    Log in for availability
    1 * 1 L
    1 * 1 L - 46,90 €
    46,90 €
    1 * 1 L
       46,90 €

  • Buffer solution
    5,00
    ±0,01
    25 °C
    Plastic bottle
    1 L
    Log in for availability
    Log in for availability
    1 * 1 L
    1 * 1 L - 49,30 €
    49,30 €
    1 * 1 L
       49,30 €

  • REGE5050
    Buffer solution
    Buffer solution
    5,00
    ±0,01
    20 °C
    Plastic bottle
    5 L
    Log in for availability
    Log in for availability
    1 * 5 L
    1 * 5 L - 141,00 €
    141,00 €
    1 * 5 L
       141,00 €

  • REGE10605
    Buffer solution
    Buffer solution
    6,00
    ±0,01
    20 °C
    Plastic bottle
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL - 31,90 €
    31,90 €
    1 * 500 mL
       31,90 €

  • Buffer solution
    6,00
    ±0,01
    25 °C
    Plastic bottle
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL - 32,90 €
    32,90 €
    1 * 500 mL
       32,90 €

  • REGE1060
    Buffer solution
    Buffer solution
    6,00
    ±0,01
    20 °C
    Plastic bottle
    1 L
    Log in for availability
    Log in for availability
    1 * 1 L
    1 * 1 L - 46,90 €
    46,90 €
    1 * 1 L
       46,90 €

  • REGE106025
    Buffer solution
    Buffer solution
    6,00
    ±0,01
    25 °C
    Plastic bottle
    1 L
    Log in for availability
    Log in for availability
    1 * 1 L
    1 * 1 L - 49,30 €
    49,30 €
    1 * 1 L
       49,30 €

  • REGE5060
    Buffer solution
    Buffer solution
    6,00
    ±0,01
    20 °C
    Plastic bottle
    5 L
    Log in for availability
    Log in for availability
    1 * 5 L
    1 * 5 L - 141,00 €
    141,00 €
    1 * 5 L
       141,00 €

  • REGE10685
    Buffer solution
    Buffer solution
    6,80
    ±0,01
    20 °C
    Plastic bottle
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL - 31,90 €
    31,90 €
    1 * 500 mL
       31,90 €

  • REGE1068525
    Buffer solution
    Buffer solution
    6,80
    ±0,01
    25 °C
    Plastic bottle
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL - 32,90 €
    32,90 €
    1 * 500 mL
       32,90 €

  • REGE1068
    Buffer solution
    Buffer solution
    6,80
    ±0,01
    20 °C
    Plastic bottle
    1 L
    Log in for availability
    Log in for availability
    1 * 1 L
    1 * 1 L - 46,90 €
    46,90 €
    1 * 1 L
       46,90 €

  • REGE106825
    Buffer solution
    Buffer solution
    6,80
    ±0,01
    25 °C
    Plastic bottle
    1 L
    Log in for availability
    Log in for availability
    1 * 1 L
    1 * 1 L - 49,30 €
    49,30 €
    1 * 1 L
       49,30 €

  • Buffer solution
    7,00
    ±0,01
    20 °C
    Plastic bottle
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL - 30,50 €
    30,50 €
    1 * 500 mL
       30,50 €

  • Buffer solution
    7,00
    ±0,01
    25 °C
    Plastic bottle
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL - 31,90 €
    31,90 €
    1 * 500 mL
       31,90 €

  • Buffer solution
    7,00
    ±0,01
    20 °C
    Plastic bottle
    1 L
    Log in for availability
    Log in for availability
    1 * 1 L
    1 * 1 L - 43,50 €
    43,50 €
    1 * 1 L
       43,50 €

  • Buffer solution
    7,00
    ±0,01
    25 °C
    Plastic bottle
    1 L
    Log in for availability
    Log in for availability
    1 * 1 L
    1 * 1 L - 46,20 €
    46,20 €
    1 * 1 L
       46,20 €

  • REGE5070
    Buffer solution
    Buffer solution
    7,00
    ±0,01
    20 °C
    Plastic bottle
    5 L
    Log in for availability
    Log in for availability
    1 * 5 L
    1 * 5 L - 127,00 €
    127,00 €
    1 * 5 L
       127,00 €

  • REGE507025
    Buffer solution
    Buffer solution
    7,00
    ±0,01
    25 °C
    Plastic bottle
    5 L
    Log in for availability
    Log in for availability
    1 * 5 L
    1 * 5 L - 134,00 €
    134,00 €
    1 * 5 L
       134,00 €

  • REGE10805
    Buffer solution
    Buffer solution
    8,00
    ±0,01
    20 °C
    Plastic bottle
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL - 31,90 €
    31,90 €
    1 * 500 mL
       31,90 €

  • Buffer solution
    8,00
    ±0,01
    25 °C
    Plastic bottle
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL - 32,90 €
    32,90 €
    1 * 500 mL
       32,90 €

  • REGE1080
    Buffer solution
    Buffer solution
    8,00
    ±0,01
    20 °C
    Plastic bottle
    1 L
    Log in for availability
    Log in for availability
    1 * 1 L
    1 * 1 L - 46,90 €
    46,90 €
    1 * 1 L
       46,90 €

  • REGE108025
    Buffer solution
    Buffer solution
    8,00
    ±0,01
    25 °C
    Plastic bottle
    1 L
    Log in for availability
    Log in for availability
    1 * 1 L
    1 * 1 L - 49,30 €
    49,30 €
    1 * 1 L
       49,30 €

  • REGE5080
    Buffer solution
    Buffer solution
    8,00
    ±0,01
    20 °C
    Plastic bottle
    5 L
    Log in for availability
    Log in for availability
    1 * 5 L
    1 * 5 L - 141,00 €
    141,00 €
    1 * 5 L
       141,00 €

  • Buffer solution
    9,00
    ±0,01
    20 °C
    Plastic bottle
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL - 30,50 €
    30,50 €
    1 * 500 mL
       30,50 €

  • Buffer solution
    9,00
    ±0,01
    25 °C
    Plastic bottle
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL - 31,90 €
    31,90 €
    1 * 500 mL
       31,90 €

  • Buffer solution
    9,00
    ±0,01
    20 °C
    Plastic bottle
    1 L
    Log in for availability
    Log in for availability
    1 * 1 L
    1 * 1 L - 43,50 €
    43,50 €
    1 * 1 L
       43,50 €

  • Buffer solution
    9,00
    ±0,01
    25 °C
    Plastic bottle
    1 L
    Log in for availability
    Log in for availability
    1 * 1 L
    1 * 1 L - 46,20 €
    46,20 €
    1 * 1 L
       46,20 €

  • REGE5090
    Buffer solution
    Buffer solution
    9,00
    ±0,01
    20 °C
    Plastic bottle
    5 L
    Log in for availability
    Log in for availability
    1 * 5 L
    1 * 5 L - 127,00 €
    127,00 €
    1 * 5 L
       127,00 €

  • Buffer solution
    10,00
    ±0,01
    20 °C
    Plastic bottle
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL - 30,50 €
    30,50 €
    1 * 500 mL
       30,50 €

  • Buffer solution
    10,00
    ±0,01
    25 °C
    Plastic bottle
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL - 31,90 €
    31,90 €
    1 * 500 mL
       31,90 €

  • REGE1100
    Buffer solution
    Buffer solution
    10,00
    ±0,01
    20 °C
    Plastic bottle
    1 L
    Log in for availability
    Log in for availability
    1 * 1 L
    1 * 1 L - 43,50 €
    43,50 €
    1 * 1 L
       43,50 €

  • Buffer solution
    10,00
    ±0,01
    25 °C
    Plastic bottle
    1 L
    Log in for availability
    Log in for availability
    1 * 1 L
    1 * 1 L - 46,20 €
    46,20 €
    1 * 1 L
       46,20 €

  • REGE5100
    Buffer solution
    Buffer solution
    10,00
    ±0,01
    20 °C
    Plastic bottle
    5 L
    Log in for availability
    Log in for availability
    1 * 5 L
    1 * 5 L - 127,00 €
    127,00 €
    1 * 5 L
       127,00 €

  • Buffer solution
    11,00
    ±0,05
    20 °C
    Plastic bottle
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL - 31,90 €
    31,90 €
    1 * 500 mL
       31,90 €

  • REGE1110525
    Buffer solution
    Buffer solution
    11,00
    ±0,05
    25 °C
    Plastic bottle
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL - 32,90 €
    32,90 €
    1 * 500 mL
       32,90 €

  • REGE1110
    Buffer solution
    Buffer solution
    11,00
    ±0,05
    20 °C
    Plastic bottle
    1 L
    Log in for availability
    Log in for availability
    1 * 1 L
    1 * 1 L - 46,90 €
    46,90 €
    1 * 1 L
       46,90 €

  • REGE111025
    Buffer solution
    Buffer solution
    11,00
    ±0,05
    25 °C
    Plastic bottle
    1 L
    Log in for availability
    Log in for availability
    1 * 1 L
    1 * 1 L - 49,30 €
    49,30 €
    1 * 1 L
       49,30 €

  • REGE11205
    Buffer solution
    Buffer solution
    12,00
    ±0,05
    20 °C
    Plastic bottle
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL - 31,90 €
    31,90 €
    1 * 500 mL
       31,90 €

  • REGE1120525
    Buffer solution
    Buffer solution
    12,00
    ±0,05
    25 °C
    Plastic bottle
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL - 32,90 €
    32,90 €
    1 * 500 mL
       32,90 €

  • REGE1120
    Buffer solution
    Buffer solution
    12,00
    ±0,05
    20 °C
    Plastic bottle
    1 L
    Log in for availability
    Log in for availability
    1 * 1 L
    1 * 1 L - 46,90 €
    46,90 €
    1 * 1 L
       46,90 €

  • Buffer solution
    12,00
    ±0,05
    25 °C
    Plastic bottle
    1 L
    Log in for availability
    Log in for availability
    1 * 1 L
    1 * 1 L - 49,30 €
    49,30 €
    1 * 1 L
       49,30 €

  • REGE5120
    Buffer solution
    Buffer solution
    12,00
    ±0,05
    20 °C
    Plastic bottle
    5 L
    Log in for availability
    Log in for availability
    1 * 5 L
    1 * 5 L - 141,00 €
    141,00 €
    1 * 5 L
       141,00 €

  • REGE512025
    Buffer solution
    Buffer solution
    12,00
    ±0,05
    25 °C
    Plastic bottle
    5 L
    Log in for availability
    Log in for availability
    1 * 5 L
    1 * 5 L - 148,00 €
    148,00 €
    1 * 5 L
       148,00 €

  • Buffer solution
    13,00
    ±0,05
    20 °C
    Plastic bottle
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL - 31,90 €
    31,90 €
    1 * 500 mL
       31,90 €

  • Buffer solution
    13,00
    ±0,05
    25 °C
    Plastic bottle
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL - 32,90 €
    32,90 €
    1 * 500 mL
       32,90 €

  • REGE1130
    Buffer solution
    Buffer solution
    13,00
    ±0,05
    20 °C
    Plastic bottle
    1 L
    Log in for availability
    Log in for availability
    1 * 1 L
    1 * 1 L - 46,90 €
    46,90 €
    1 * 1 L
       46,90 €

  • REGE113025
    Buffer solution
    Buffer solution
    13,00
    ±0,05
    25 °C
    Plastic bottle
    1 L
    Log in for availability
    Log in for availability
    1 * 1 L
    1 * 1 L - 49,30 €
    49,30 €
    1 * 1 L
       49,30 €

  • REGE5130
    Buffer solution
    Buffer solution
    13,00
    ±0,05
    20 °C
    Plastic bottle
    5 L
    Log in for availability
    Log in for availability
    1 * 5 L
    1 * 5 L - 141,00 €
    141,00 €
    1 * 5 L
       141,00 €

This product has been blocked by your organization. Please contact your purchasing department for more information

pH buffer standards DIN 19266

Product Information

  • pH value
    pH accuracy
    Colour
    Reference temperature
    Pack type
    Pk
    Availability
    Price
  • REGE101679
    Buffer standard DIN 19266
    Buffer standard DIN 19266
    1,679
    -
    25 °C
    Plastic bottle
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL - 41,70 €
    41,70 €
    1 * 500 mL
       41,70 €

  • REGE103776
    Buffer standard DIN 19266
    Buffer standard DIN 19266
    3,776
    -
    25 °C
    Plastic bottle
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL - 41,70 €
    41,70 €
    1 * 500 mL
       41,70 €

  • REGE104005
    Buffer standard DIN 19266
    Buffer standard DIN 19266
    4,005
    -
    25 °C
    Plastic bottle
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL - 41,70 €
    41,70 €
    1 * 500 mL
       41,70 €

  • REGE10687
    Buffer standard DIN 19266
    Buffer standard DIN 19266
    6,865
    -
    25 °C
    Plastic bottle
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL - 41,70 €
    41,70 €
    1 * 500 mL
       41,70 €

  • REGE109180
    Buffer standard DIN 19266
    Buffer standard DIN 19266
    9,180
    -
    25 °C
    Plastic bottle
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL - 41,70 €
    41,70 €
    1 * 500 mL
       41,70 €

  • REGE110012
    Buffer standard DIN 19266
    Buffer standard DIN 19266
    10,012
    -
    25 °C
    Plastic bottle
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL - 41,70 €
    41,70 €
    1 * 500 mL
       41,70 €

  • REGE112454
    Buffer standard DIN 19266
    Buffer standard DIN 19266
    12,454
    -
    25 °C
    Plastic bottle
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL - 41,70 €
    41,70 €
    1 * 500 mL
       41,70 €

High-resolution buffers

Product Information

  • pH value
    pH accuracy
    Colour
    Reference temperature
    Pack type
    Pk
    Availability
    Price
  • REGEH70525
    High-resolution buffer
    High-resolution buffer
    7,000
    -
    25 °C
    Plastic bottle
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL - 40,60 €
    40,60 €
    1 * 500 mL
       40,60 €

  • REGEH70525C
    High Resolution Colour Coded Buffer
    High Resolution Colour Coded Buffer
    7,000
    -
    Yellow
    25 °C
    Plastic bottle
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL - 42,10 €
    42,10 €
    1 * 500 mL
       42,10 €