Orders LinkContactUsLinkComponent
Multiple adapters
Multiple adapters
Supplier:   Lenz Laborglas
Multiple adapters
Supplier:  Lenz Laborglas
Certificates
Multiple adapters
Multiple adapters
5 Options Available Below
Product Details & Documents
DURAN® Borosilicate glass 3.3
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
With 3 NS sockets and 1 NS cone, with two parallel necks and one angled neck (45°).
Documents
Product Family Options
Product Information
  • Cone size
    Socket size
    Availability
    Price
  • 14/23 NS
    14/23 NS
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 59,80 €
    59,80 €
    1 * 1 ST
       59,80 €
  • 29/32 NS
    19/26 NS
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 65,20 €
    65,20 €
    1 * 1 ST
       65,20 €
  • 29/32 NS
    29/32 NS
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 82,80 €
    82,80 €
    1 * 1 ST
       82,80 €
  • 24/29 NS
    19/26 NS
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 65,20 €
    65,20 €
    1 * 1 ST
       65,20 €
  • 24/29 NS
    24/29 NS
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 69,90 €
    69,90 €
    1 * 1 ST
       69,90 €