Order Entry
Austria
ContactUsLinkComponent
2,2-Dimethylbutane, Supelco®
2,2-Dimethylbutane, Supelco®
Catalog # 39730-5ML
Supplier:  Merck
CAS Number:  
undefined
2,2-Dimethylbutane, Supelco®
Catalog # 39730-5ML
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    5 mL
  • Seq:
    2210
  • Environmentally Preferable:

Specifications

About this item

Synonyms: Neohexane
 :
 :86.18 g/mol
EINECS:200-906-8
UN:1208
 :GHS02
 :GHS08
 :GHS07
 :GHS09
Storage Temperature:Ambient
CAS Number:75-83-2
Formula:C₆H₁₄
MDL Number:MFCD00009321
Boiling Pt:49.7
Melting Pt:–100 °C
Density:0.649 g/cm³ (20 °C)
ADR:3 II
 :M-Clarity™ quality level = MQ100