Order Entry
Austria
ContactUsLinkComponent
2,2,4-Trimethylpentane
Flammable
Health
Irritant
Environment
2,2,4-Trimethylpentane
Catalog # PROOHDF01
CAS Number:  
undefined
Flammable
Health
Irritant
Environment
2,2,4-Trimethylpentane
Catalog # PROOHDF01
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    10 mL
  • Seq:
    2275

Specifications

About this item

 :
 :114.23 g/mol
Merck Index:13,05212
Boiling Pt:99.2 °C (1013 hPa)
Melting Pt:–107 °C
Density:0.692 g/cm³ (20 °C)
Flash Pt:–12 °C
Storage Temperature:Ambient
EINECS:208-759-1
UN:1262
ADR:3,II
 :GHS02
 :GHS08
 :GHS07
 :GHS09
CAS Number:540-84-1
Formula:(CH₃)₂CHCH₂C(CH₃)₃
MDL Number:MFCD00008943