Disposable earplugs, E-A-RSoft™ FX
Supplier: 3M
Certificates
About this item
This foam earplug with tapered bell shape and super-soft foam is easy-to-fit and seals effectively to provide the highest NRR available in the industry. Custom polyurethane foam formulation provides slow recovery and extreme softness for optimum comfort.
- Flange shape serves as an effective insertion device when rolled down
- Easy to fit into the ear canal
- Available with or without cord
- Compatible with other PPE
SNR: 39 dB
Frequently Bought Together