Order Entry
Austria
ContactUsLinkComponent
 
2,2,4-Trimethylpentane ≥99% (by GC), CHROMASOLV™ for pesticide residue analysis, Riedel-de Haën™
 
undefined
2,2,4-Trimethylpentane ≥99% (by GC), CHROMASOLV™ for pesticide residue analysis, Riedel-de Haën™

 

  • Vapor Density:
    3.9 (vs air)
  • Assay (GC):
    Min. 99 %
  • Non-volatile matter:
    Max. 0.0005 %
  • Water (Karl Fischer):
    Max. 0.01 %
  • Suitability for residue analysis:
    Max. 5 ng/l
  • Determination of P/N:
    ≤5 ng/L
  • Autoignition Temperature:
    745 °F
  • Explosion Limit:
    6 %
  • Vapor Pressure:
    41 mmHg ( 21 °C)

 

 

Frequently Bought Together