Order Entry
Austria
ContactUsLinkComponent
2,2,4-Trimethylpentane ≥99.8% (by GC, corrected for water content) for HPLC, J.T.Baker®
2,2,4-Trimethylpentane ≥99.8% (by GC, corrected for water content) for HPLC, J.T.Baker®
Supplier:  Avantor
Certificates
undefined
2,2,4-Trimethylpentane ≥99.8% (by GC, corrected for water content) for HPLC, J.T.Baker®
Supplier:  Avantor
Certificates

Specification Test Results

  • For Laboratory,Research,or Manufacturing Use:
    min. 99.8 %
  • Ultraviolet Absorbance (1.00-cm cell vs. water) - 400-280 nm:
    ≤ 0.01
  • Ultraviolet Absorbance (1.00-cm cell vs. water) - 254 nm:
    ≤ 0.015
  • Ultraviolet Absorbance (1.00-cm cell vs. water) - 225 nm:
    ≤ 0.10
  • Ultraviolet Absorbance (1.00-cm cell vs. water) - UV Cut-off, nm:
    ≤ 205
  • Fluorescence Trace Impurities, measured as Quinine Base - at 450 nm Emission:
    ≤ 0.3 ppb
  • Fluorescence Trace Impurities, measured as Quinine Base - at Emission Maximum for Impurities:
    ≤ 1.0 ppb
  • Assay (by GC, corrected for water):
    ≥ 99.8 %
  • Residue after Evaporation:
    ≤ 2.0 ppm
  • Titrable Acid (µeq/g):
    ≤ 0.3
  • Water (by KF, coulometric):
    ≤ 100 ppm

Specification Test Results

Related Information

Frequently Bought Together