Order Entry
Austria
ContactUsLinkComponent
2,2,4-Trimethylpentane, BAKER ANALYZED® ACS, J.T.Baker®
2,2,4-Trimethylpentane, BAKER ANALYZED® ACS, J.T.Baker®
Supplier:  Avantor
Certificates
undefined
2,2,4-Trimethylpentane, BAKER ANALYZED® ACS, J.T.Baker®
Supplier:  Avantor
Certificates

Specification Test Results

  • Assay (by GC):
    min. 99.0 %
  • Color (APHA):
    max. 10
  • Residue after Evaporation:
    max. 5 ppm
  • Sulfur Compounds (as S):
    max. 0.005 %
  • Water Soluble Titrable Acid (meq/g):
    max. 0.0003
  • at 210 nm:
    max. 1.0
  • at 220 nm:
    max. 0.20
  • at 230 nm:
    max. 0.10
  • at 240 nm:
    max. 0.04
  • at 250 - 400nm:
    max. 0.01

Specification Test Results

Related Information

Frequently Bought Together