2,2,4-Trimethylpentane, extra dry 99.5%, AcroSeal®
:540-84-1
:
:(CH₃)₂CHCH₂C(CH₃)₃
:MFCD00008943
:114.23 g/mol
:13,05212
:98…99 °C (760 mmHg)
:–107 °C
:0.69 g/cm³
:–12 °C
:208-759-1
:1262
:3,II
:GHS07
:GHS08
:GHS09
:GHS02
:Ambient
- Appearance:Clear colorless Liquid
- Infrared spectrum:Conforms
- Water:≤0.005 % (Coulometric)
- GC:≥99.4 %
- Residue after evaporation:≤0.0005 %
Frequently Bought Together