2,2,4-Trimethylpentane 99.5%, ECD tested for pesticide analysis for residue analysis
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:540-84-1
:
Formula:(CH₃)₂CHCH₂C(CH₃)₃
MDL Number:MFCD00008943
:114.23 g/mol
Merck Index:13,05212
Boiling Pt:99.2 °C (1013 hPa)
Melting Pt:–107 °C
Density:0.69 g/cm³
Flash Pt:–12 °C
EINECS:208-759-1
UN:1262
ADR:3,II
:GHS07
:GHS08
:GHS09
:GHS02
Storage Temperature:Ambient
Specification Test Results
- Appearance:Clear liquid
- Water:≤0.01 % (Coulometric)
- GC:≥99.5 %
- Color scale:≤10 APHA
- GC/ECD (as lindane):≤5 ng/l
- Residue after evaporation:≤0.0005 %
- Acidity (CH3COOH):≤0.003 %
Specification Test Results
Frequently Bought Together