Order Entry
Austria
ContactUsLinkComponent
2,2,4-Trimethylpentane 99+%, pure
Irritant
Health
Environment
Flammable
2,2,4-Trimethylpentane 99+%, pure
Certificates
undefined
Irritant
Health
Environment
Flammable
2,2,4-Trimethylpentane 99+%, pure

Specification Test Results

  • Appearance:
    Clear colorless Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.3900 to 1.3920 (20°C, 589 nm)
  • GC:
    ≥99.0 %

Specification Test Results

Related Information

Frequently Bought Together