Order Entry
Austria
ContactUsLinkComponent
2,2,4-Trimethylhexane, Sigma-Aldrich®
2,2,4-Trimethylhexane, Sigma-Aldrich®
Catalog # SIAL92470-5ML
Supplier:  Merck
CAS Number:  
2,2,4-Trimethylhexane, Sigma-Aldrich®
Catalog # SIAL92470-5ML
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    5 mL
  • Seq:
    2210
  • Environmentally Preferable:

Specifications

About this item

 :
 :128.26 g/mol
Melting Pt:–120 °C
EINECS:240-807-7
UN:1920
 :GHS02
 :GHS08
Storage Temperature:Ambient
CAS Number:16747-26-5
Formula:C₉H₂₀
MDL Number:MFCD00039928
Boiling Pt:127
Density:0.711 g/cm³ (20 °C)
ADR:3 III