Order Entry
Austria
ContactUsLinkComponent
Magnetic stirring bar retrievers, RSE, PTFE
Magnetic stirring bar retrievers, RSE, PTFE
Catalog # 442-0902
Supplier:  IKA
Magnetic stirring bar retrievers, RSE, PTFE
Catalog # 442-0902
Supplier:  IKA

Specifications

  • Ø:
    8 mm
  • Length:
    350 mm
  • Pk:
    1

Specifications

About this item

These PTFE-coated retriever used for removing all stirring bars upto 80 mm in length.

  • Weight: 0,048 kg