Order Entry
Austria
ContactUsLinkComponent
2-Chloro-2-methylpropane 99%
2-Chloro-2-methylpropane 99%
Certificates
undefined
2-Chloro-2-methylpropane 99%
Certificates

Specifications

  • Environmentally Preferable:

Specifications

Specification Test Results

  • Appearance:
    Clear liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.3838 to 1.3858 (20°C, 589 nm)
  • GC:
    ≥98.5 %
  • Color scale:
    ≤15 APHA

Specification Test Results

Related Information

Frequently Bought Together