Joint clamps for conical ground joints, Safety Clip
Clamps made of POM, for firm and secure fitting of conical ground joints.
- Will not scratch glass surfaces
- Suitable for use with sleeves with grip collar
- Outstanding chemical resistance
- Operating temperature up to 150 °C
Frequently Bought Together