5-Methyl-2-hexanone ≥99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: Isoamyl methyl ketone, Methyl isoamyl ketone
CAS Number:110-12-3
:
Formula:C₇H₁₄O
MDL Number:MFCD00008950
:114.19 g/mol
Boiling Pt:144…145 °C
Melting Pt:–74 °C
Density:0.812
Flash Pt:41 °C (105 °F)
Storage Temperature:Ambient
EINECS:203-737-8
UN:2302
ADR:3,III
:GHS02
:GHS07
Specifications
- Environmentally Preferable:
Specifications
Related Information
Frequently Bought Together