Order Entry
Austria
ContactUsLinkComponent
5-Methyl-2-hexanone 99%
5-Methyl-2-hexanone 99%
Certificates
undefined
5-Methyl-2-hexanone 99%

Specifications

  • Environmentally Preferable:

Specifications

Specification Test Results

  • Appearance:
    Clear colorless Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.4060 to 1.4090 (20°C, 589 nm)
  • GC:
    ≥98.5 %
  • Specific optical rotation:
    +16° to +23° (20°C, 589 nm) (c=1 in H2O)

Specification Test Results

Related Information

Frequently Bought Together