Order Entry
Austria
ContactUsLinkComponent
 
4-Methyl-2-pentanone, EMSURE® ACS, Reag. Ph. Eur. for analysis, Supelco®
 :  Merck
    
 
undefined
4-Methyl-2-pentanone, EMSURE® ACS, Reag. Ph. Eur. for analysis, Supelco®
 :  Merck
    
 
 
 

 

  • Environmentally Preferable:
    Safer Human and Environmental Health
  • Environmental certification:
    Chem21 Solvent Selection Guide recommended by default and after discussion

 

 

  • Solubility:
    18 - 20 g/l (20 °C)
  • Melting Point:
    -84 °C
  • Molar Mass:
    100.16 g/mol
  • Boiling Point:
    116 - 118 °C (1013 hPa)
  • Vapor Pressure:
    20 hPa (20 °C)
  • Flash Point:
    14 °C
  • Refractive Index:
    1.3962 (20 °C)
  • Explosion Limit:
    1.2 - 8.0 %(V)
  • Density:
    0.80 g/cm3 (20 °C)
  • pH:
    (H2O, 20 °C) neutral
  • Ignition Point:
    460 °C
  • Evaporation number:
    10

 

Frequently Bought Together