Order Entry
Austria
Orders LinkContactUsLinkComponent
3-Methyl-2-pentanone ≥98%

3-Methyl-2-pentanone ≥98%

3-Methyl-2-pentanone ≥98%
Certificates
3-Methyl-2-pentanone ≥98%
3-Methyl-2-pentanone ≥98%

3 Options Available Below

Product Details & Documents

Purity: 98+% CAS No.: 565-61-7 Molecular Formula: C6H12O Formula Weight: 100.16 Flash Point: 12°(53°F) Refractive Index: 1.4 00 Boiling Point: 118-119° Beilstein Reference: 635791 UN No.: 1224 MDL No.: MFCD00009336

  • CAS Number:565-61-7
  • Formula:C₆H₁₂O
  • MDL Number:MFCD00009336
  • Molecular Weight:100.16 g/mol
  • Boiling Pt:118…119 °C
  • Density:0.815
  • Flash Pt:12 °C (53 °F)
  • Storage Temperature:Ambient
  • EINECS:209-282-1
  • UN:1224
  • ADR:3,II
Product Family Options

Product Information

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.