
Isobutyraldehyde
Catalog # EHERCA14395000
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 mL
- Seq:2070
Specifications
About this item
:
:72.11 g/mol
Merck Index:13,05171
Boiling Pt:64 °C (1013 hPa)
Melting Pt:–65 °C
Density:0.794 g/cm³ (20 °C)
Flash Pt:–24 °C
Storage Temperature:Ambient
EINECS:201-149-6
UN:2045
ADR:3,II
:GHS02
CAS Number:78-84-2
Formula:C₄H₈O
MDL Number:MFCD00006980