Order Entry
Austria
Orders LinkContactUsLinkComponent
 

n-Pentane ≥95%, TECHNICAL

Flammable
Health
Irritant
Environment
Supplier:   VWR Chemicals
n-Pentane ≥95%, TECHNICAL
Supplier:  VWR Chemicals
Certificates
n-Pentane ≥95%, TECHNICAL

4 Options Available Below

Product Details & Documents
Synonyms:  Pentane
  • CAS Number:109-66-0
  • Formula:H₃C(CH₂)₃CH₃
  • MDL Number:MFCD00009498
  • Molecular Weight:72.15 g/mol
  • Merck Index:13,07193
  • Boiling Pt:35…36 °C (1013 hPa)
  • Melting Pt:–130 °C
  • Density:0.63 g/cm³ (20 °C)
  • Flash Pt:–40 °C
  • Storage Temperature:Ambient
  • EINECS:203-692-4
  • UN:1265
  • ADR:3,II
  • REACH:01-2119459286-30
Specifications for Product Family

Specification Test Results

  • Assay:
    Min. 95 %
  • Boiling point:
    35 - 37 °C
Product Family Options

Product Information