n-Pentane ≥99.9% (by GC), Suprasolv® hypergrade for organic trace analysis, Supelco®
Supplier: Merck
Certificates
About this item
CAS Number:109-66-0
:
Formula:H₃C(CH₂)₃CH₃
MDL Number:MFCD00009498
:72.15 g/mol
Merck Index:13,07193
Boiling Pt:35…36 °C (1013 hPa)
Melting Pt:–130 °C
Density:0.63 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:203-692-4
UN:1265
ADR:3,II
:M-Clarity™ quality level = MQ100
Flash Pt:–48 °C
REACH:01-2119459286-30
:GHS02
:GHS08
:GHS07
:GHS09
Specification Test Results
- Purity (GC):≥ 99.9%
- Identity (IR):Conforms
- Evaporation Residue:≤3.0 mg/L
- Water:≤0.01%
- Color:≤10 Hazen
- GC/ECD (Retention Range 1,2,4-tri- Chlorobenzene to Decachlorobiphenyle, Individual Signals (Lindane Standard)):≤2 pg/ml
- GC/ECD (Retention Range Dichloromethane to 1,2,4-Trichlorobenzene Individual Signals (Tetrachloromethane)):≤1 ng/ml
- GC/FID (Retention Range n-Undecane to n-Tetracontane, Individual Signals (n-Tetradecane Standard)):≤2 ng/ml
- GC/MSD (Retention Range n-Undecane to n-Tetracontane, Scaning area 30 - 600 amu, Individual Signals (n- Tetradecane Standard)):≤2 ng/ml
Specification Test Results
Related Information
Frequently Bought Together