Order Entry
Austria
ContactUsLinkComponent
 
n-Pentane, EMSURE® for analysis, Supelco®
 :  Merck
 
undefined
n-Pentane, EMSURE® for analysis, Supelco®
 :  Merck
 
 
 

 

  • Solubility:
    0.04 g/l (25 °C)
  • Melting Point:
    -130 °C
  • Molar Mass:
    72.15 g/mol
  • Boiling Point:
    36 °C (1013 hPa)
  • Vapor Pressure:
    570 hPa (20 °C)
  • Flash Point:
    -48 °C (refers to pure substance)
  • Refractive Index:
    1.358
  • Explosion Limit:
    1.4 - 8 %(V)
  • Density:
    0.63 g/cm3 (20 °C)
  • Ignition Point:
    285 °C

 

Frequently Bought Together