Orders LinkContactUsLinkComponent
Cover spectacles, 2800
Supplier:   3M
Cover spectacles, 2800
Supplier:  3M
Certificates
Cover spectacles, 2800
Cover spectacles, 2800
3 Options Available Below
Product Details & Documents
These lightweight cover glasses (OTG) are designed to fit over most prescription spectacles with minimal interference. Available in three lens options. The green lens offers protection against infrared radiation arising from certain gas welding, gas cutting and brazing applications (IR shade 5).
  • Strong polycarbonate lens with anti-scratch coating
  • Adjustable temple length (four positions)
  • Inclinable lens for easy adjustment and maximum comfort
  • Soft low profile temples for comfort and minimum interaction with prescription spectacles
Weight: 38 g
 : EN 166, EN 170, EN 171 (for IR), FT 1
Product Family Options
Product Information
  • Type
    Colour
    Lenses
    Availability
    Price
  • Anti-UV (2C - 1,2), anti-scratch, anti-fog
    Blue
    Clear PC
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 12,00 €
    12,00 €
    1 * 1 ST
       12,00 €
  • Anti-UV (2 - 1,2), anti-scratch, anti-fog
    Blue
    Yellow PC
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 12,00 €
    12,00 €
    1 * 1 ST
       12,00 €
  • Anti-UV, IR (4-5), anti-scratch, anti-fog
    Blue
    Green PC
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 20,70 €
    20,70 €
    1 * 1 ST
       20,70 €