Burettes, pellet pattern, without intermediate stopcock, SILBERBRAND
Supplier: Brand
Jump to:
About this item
Amber borosilicate glass 5.4, class B.
- Calibrated to deliver 'TD, Ex'
- Short marks at major graduations, for 25 ml reduced distance between subdivision marks
- Automatic zeroing, intermediate stopcock for recirculating residual liquid
25 ml: Error limits according to class B, DIN EN ISO 385
50 ml: DIN EN ISO 385, class B
*Total height includes height of reservoir bottle.
2 Options Available Below
Product Details & Documents
Amber borosilicate glass 5.4, class B.
- Calibrated to deliver 'TD, Ex'
- Short marks at major graduations, for 25 ml reduced distance between subdivision marks
- Automatic zeroing, intermediate stopcock for recirculating residual liquid
25 ml: Error limits according to class B, DIN EN ISO 385
50 ml: DIN EN ISO 385, class B
*Total height includes height of reservoir bottle.
Documents
Brand Volumetric Measurement in the Laboratory
Product Literature
Product Family Options
Burette with titration stopcock with PTFE key without reservoir bottle
Product Information
Accessories
Product Information
- Catalog #CapacityAvailabilityPrice
- Reservoir bottle, amber glass
Product Information
- Catalog #CapacityAvailabilityPrice
- Ball set with net, black, size 5, natural rubber/PVCBall set with net, black, size 5, natural rubber/PVC612-9953102Log in for availability1 * 1 ST - 107,00 €
Product Information
- Catalog #AvailabilityPrice
- Single bulb rubber bellows, orange coloured, with valve and tubingSingle bulb rubber bellows, orange coloured, with valve and tubingBRND23400Log in for availability1 * 1 ST - 16,10 €