Order Entry
Austria
ContactUsLinkComponent
DL-α-Tocopherol
DL-α-Tocopherol
Catalog # 47783.
Supplier:  Merck
CAS Number:  
undefined
DL-α-Tocopherol
Catalog # 47783.
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1
  • Pk Info:
    100 mg

Specifications

About this item

 :
 :430.71 g/mol
EINECS:233-466-0
CAS Number:10191-41-0
 :1406-66-2
Formula:C₂₉H₅₀O₂
MDL Number:MFCD00072051