β-Propiolactone
Artikelnr. MOLE68711089-1ML
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 mL
- Seq:2070
Specifications
About this item
:
:72.06 g/mol
Boiling Pt:155 °C (1013 hPa)
Melting Pt:–31.2 °C
Density:1.145 g/cm³ (20 °C)
EINECS:200-340-1
UN:3382
ADR:6.1,I
CAS Number:57-57-8
Formula:C₃H₄O₂
MDL Number:MFCD00005169