Order Entry
Austria
ContactUsLinkComponent
n-Octane 99+%, extra pure
Irritant
Health
Environment
Flammable
n-Octane 99+%, extra pure
Certificates
undefined
Irritant
Health
Environment
Flammable
n-Octane 99+%, extra pure

Specification Test Results

  • Appearance:
    Clear colorless Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.3965 to 1.3985 (20°C, 589 nm)
  • Density:
    (15°C) 0.706 to 0.710 kg/l
  • GC:
    ≥99.0 %
  • Residue after evaporation:
    ≤0.05 %

Specification Test Results

Related Information

Frequently Bought Together