Cell culture tubes, Nunc™
Catalog # 392-0334
Supplier: Thermo Fisher Scientific
Specifications
- Ø:17,5 mm
- Capacity:14 ml
- Description:Round bottom tube, with dual position polyethylene cap, sterile
- Height:95 mm
- Pk:500
Specifications
About this item
Highly certified tubes for critical cellular applications.
- Optimised gas exchange with dual position vent/close cap
- Off-gas volatile organic compounds easily with breathable packaging
- Certificates of conformity available for every lot
- Non-pyrogenic
- For research use only
Mouse Embryo Assay (MEA) tested. Produced under ISO 13485 with full lot traceability. USP Class VI tested. Certified.