Order Entry
Austria
Orders LinkContactUsLinkComponent
Ashless filter aids, Whatman™

Ashless filter aids, Whatman™

Ashless clippings
Catalog # WHAT1703-050
Ashless filter aids, Whatman™
Catalog # WHAT1703-050
Ashless filter aids, Whatman™
Ashless filter aids, Whatman™

Ashless clippings

Price
102,00 €
Log in for availability
1 * 500 g
Product Details & Documents

Ashless filter aids enhance filtration speed by coagulating precipitates or suspensions to form a thick retentive ‘prefilter’ layer on top of normal filter paper.

  • Easily dispersible
  • Powder is of a purity similar to that of ashless quantitative papers
  • Maximum ash content is 0,015%

Delivery information: Supplied with two ended scoop for measuring 0,50 or 2,5 g quantities.

Specifications
  • Description:
    Ashless clippings
  • Cat. No.:
    WHAT1703-050
  • Ash content:
    0,015%
  • Weight:
    500 g
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.