Digital Solutions
Applications
Support
About
Austria
ContactUsLinkComponent
Stopcocks
Replacement stopcock for aspirator bottles
Replacement stopcock for aspirator bottles
BRND131100
:
Brand
Replacement stopcock for aspirator bottles
BRND131100
:
Brand
Description:
Spare stopcock, 19.05 mm, for aspirator bottles, with PP adapter and 2 FKM seals
Pk:
1