n-Hexatriacontane
Catalog # EHERC14199000
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:0,1 g
- Seq:3020
Specifications
About this item
:
:506.98 g/mol
Boiling Pt:497 °C (1013 hPa)
Melting Pt:75.5 °C
Density:0.7803 g/cm³ (80 °C)
Storage Temperature:Ambient
EINECS:211-127-8
CAS Number:630-06-8
Formula:H₃C(CH₂)₃₄CH₃
MDL Number:MFCD00009413